3-Chloropropyldimethoxymethylsilane 95%

Supplier: Ambeed
Danger

Synonyms: 3-(Methyldimethoxysilyl)propyl Chloride, (3-Chloropropyl)dimethoxy(methyl)silane

A856275-25G A856275-5G
77281-730EA 13.39 USD
77281-730 77281-732
3-Chloropropyldimethoxymethylsilane 95%
3-Chloropropyldimethoxymethylsilane

Precaución: for Research Use Only

Formula: C₆H₁₅ClO₂Si
MW: 182.72 g/mol
MDL Number: MFCD00053912
CAS Number: 18171-19-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR