2-Acetoxyisobutyroyl chloride 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
α-Acetoxyisobutyryl chloride,2-Acetoxy-2-methylpropionyl chloride, 1-Chloro-2-methyl-1-oxopropan-2-yl acetate, 2-Acetoxyisobutyryl Chloride, Chlorocarbonyl-1-methylethyl acetate
A856058-10G
A856058-25G
A856058-5G
77256-338EA
22
USD
77256-338
77256-340
77256-342
2-Acetoxyisobutyroyl chloride 95%
2-Acetoxyisobutyroyl chloride
Precaución: for Research Use Only
Formula:
C₆H₉ClO₃ MW: 164.59 g/mol |
MDL Number:
MFCD00000708 CAS Number: 40635-66-3 UN: 3265 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...