Ethyl 3-hydroxy-5-oxocyclohex-3-enecarboxylate 97%

Supplier: Ambeed
Warning

A855912-100MG A855912-50MG
77268-606EA 247.2 USD
77268-606 77268-608
Ethyl 3-hydroxy-5-oxocyclohex-3-enecarboxylate 97%
Ethyl 3-hydroxy-5-oxocyclohex-3-enecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂O₄
MW: 184.18 g/mol
MDL Number: MFCD19441209
CAS Number: 88805-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR