N-Boc-PEG5-alcohol 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (14-hydroxy-3,6,9,12-tetraoxatetradecyl)carbamate

A855404-100MG A855404-1G A855404-250MG A855404-5G
77268-594EA 31.93 USD
77268-594 77268-596 77268-598 77268-600
N-Boc-PEG5-alcohol 97%
N-Boc-PEG5-alcohol

Precaución: for Research Use Only

Formula: C₁₅H₃₁NO₇
MW: 337.409 g/mol
MDL Number: MFCD27635183
CAS Number: 1404111-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR