2-Ethylaminoethanol 95%

Supplier: Ambeed
Danger

Synonyms: N-Ethyl-2-aminoethanol, N-Ethyl-2-ethanolamin, 2-(Ethylamino)ethan-1-ol

A854916-25G A854916-5G
77256-328EA 8 USD
77256-328 77256-330
2-Ethylaminoethanol 95%
2-Ethylaminoethanol

Precaución: for Research Use Only

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 167
Melting Pt: -9
MDL Number: MFCD00002841
CAS Number: 110-73-6
UN: 2922
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR