3-Acetylbenzophenone 98%

Supplier: Ambeed
Danger

Synonyms: 1-(3-Benzoylphenyl)ethan-1-one, 1-(3-Benzoylphenyl)ethanone

A853283-1G A853283-250MG A853283-5G A853283-100MG A853283-25G
77281-716EA 447 USD
77281-716 77281-718 77281-720 77618-388 77749-660
3-Acetylbenzophenone 98%
3-Acetylbenzophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₂
MDL Number: MFCD01662280
CAS Number: 66067-44-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR