Phenyl-3-chloropropionate 95%

Supplier: Ambeed
Warning

Synonyms: Phenyl 3-chloropropanoate, 3-Chloropropionic Acid Phenyl Ester

A853040-100G A853040-25G
77281-712EA 162 USD
77281-712 77281-714
Phenyl-3-chloropropionate 95%
Phenyl-3-chloropropionate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD00045298
CAS Number: 24552-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR