4,4'-Oxybis(benzenesulfonyl Chloride) 98%

Supplier: Ambeed
Danger

Synonyms: Bis(4-chlorosulphonylphenyl) Ether

A852663-25G A852663-5G
77294-008EA 19.57 USD
77294-008 77294-010
4,4'-Oxybis(benzenesulfonyl Chloride) 98%
4,4'-Oxybis(benzenesulfonyl Chloride)

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂O₅S₂
MW: 367.21 g/mol
MDL Number: MFCD00024884
CAS Number: 121-63-1
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR