4,6-Dichloro-7-(phenylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A851764-100MG A851764-250MG A851764-50MG
77244-508EA 155.53 USD
77244-508 77244-510 77244-512
4,6-Dichloro-7-(phenylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine 97%
4,6-Dichloro-7-(phenylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₂H₇Cl₂N₃O₂S
MDL Number: MFCD20487117
CAS Number: 1346447-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR