2-(2,4,6-Trichlorophenyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate 97%
Supplier: Ambeed
|
Warning
|
A850942-100MG
A850942-1G
A850942-250MG
77256-314EA
55.62
USD
77256-314
77256-316
77256-318
2-(2,4,6-Trichlorophenyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate 97%
2-(2,4,6-Trichlorophenyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₁₁H₉BCl₃F₄N₃ MW: 376.37 g/mol |
MDL Number:
MFCD30489139 CAS Number: 1353251-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...