(Dimethanesulfinylidene)amine 95%

Supplier: Ambeed
Warning

Synonyms: Dimethylsulphoximine, (S-Methylsulphonimidoyl)methane

A850682-100MG A850682-1G A850682-250MG A850682-25G A850682-5G
77268-584EA 5.15 USD
77268-584 77268-586 77268-588 77268-590 77268-592
(Dimethanesulfinylidene)amine 95%
(Dimethanesulfinylidene)amine

Precaución: for Research Use Only

Formula: C₂H₇NOS
MW: 93.15 g/mol
MDL Number: MFCD02656633
CAS Number: 1520-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR