5-Ethylpyridin-3-amine 97%

Supplier: Ambeed
Warning

A850454-100MG A850454-250MG
77268-580EA 133.9 USD
77268-580 77268-582
5-Ethylpyridin-3-amine 97%
5-Ethylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.16 g/mol
MDL Number: MFCD19543983
CAS Number: 1207529-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR