(S)-5-Benzyl-2-phenyl-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate 98%

Supplier: Ambeed
Danger

A850261-100MG A850261-1G
77256-310EA 59 USD
77256-310 77256-312
(S)-5-Benzyl-2-phenyl-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate 98%
(S)-5-Benzyl-2-phenyl-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₈H₁₈BF₄N₃O
MDL Number: MFCD28167845
CAS Number: 464174-87-6
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR