Methylmaleic hydrazide 97%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dihydroxy-4-methylpyridazine, 4-Methyl-3,6-pyridazinediol, 1,2-Dihydro-4-methyl-3,6-pyridazinedione, Methylmaleic Acid Hydrazide

A848374-100G A848374-10G A848374-1G A848374-25G A848374-500G A848374-5G
77293-986EA 106 USD
77293-986 77293-988 77293-990 77293-992 77293-994 77293-996
Methylmaleic hydrazide 97%
Methylmaleic hydrazide

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MDL Number: MFCD00067746
CAS Number: 5754-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR