3-(4-Chlorobenzoyl)propionic acid 98%

Supplier: Ambeed
Warning

A848090-100G A848090-1G A848090-25G A848090-5G
77281-700EA 162 USD
77281-700 77281-702 77281-704 77281-706
3-(4-Chlorobenzoyl)propionic acid 98%
3-(4-Chlorobenzoyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
MDL Number: MFCD00002794
CAS Number: 3984-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR