2-Ethyl-1,3-cyclopentanedione 98%

Supplier: Ambeed
Warning

A847496-100G A847496-500G A847496-5G
77268-568EA 80.34 USD
77268-568 77268-570 77268-572
2-Ethyl-1,3-cyclopentanedione 98%
2-Ethyl-1,3-cyclopentanedione

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
MDL Number: MFCD00044191
CAS Number: 823-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR