6-(Dimethylamino)pyrimidine-2,4(1H,3H)-dione 97%
Supplier: Ambeed
|
Warning
|
A846993-100MG
A846993-250MG
77256-288EA
50.47
USD
77256-288
77256-290
6-(Dimethylamino)pyrimidine-2,4(1H,3H)-dione 97%
6-(Dimethylamino)pyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only
Formula:
C₆H₉N₃O₂ MW: 155.15 g/mol |
CAS Number:
4805-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...