(S)-6-Acetamido-2-aminohexanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Nε-Acetyl-L-lysine HCl

A846079-25G A846079-5G
77244-486EA 171 USD
77244-486 77747-612
(S)-6-Acetamido-2-aminohexanoic acid hydrochloride 97%
(S)-6-Acetamido-2-aminohexanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₇ClN₂O₃
MW: 224.68 g/mol
MDL Number: MFCD09752164
CAS Number: 1820580-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR