5-Bromo-1H-pyrazole-4-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A844409-100MG
A844409-250MG
A844409-1G
A844409-5G
77244-478EA
54.59
USD
77244-478
77244-480
77573-686
77747-610
5-Bromo-1H-pyrazole-4-carbonitrile 97%
5-Bromo-1H-pyrazole-4-carbonitrile
Precaución: for Research Use Only
Formula:
C₄H₂BrN₃ MW: 171.98 g/mol |
MDL Number:
MFCD26383643 CAS Number: 1703808-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...