5-Iodo-2-methyl-4-nitro-1H-imidazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-2-methyl-5-nitro-1H-imidazole

A844215-1G A844215-250MG A844215-5G
77293-980EA 39 USD
77293-980 77293-982 77293-984
5-Iodo-2-methyl-4-nitro-1H-imidazole 95%
5-Iodo-2-methyl-4-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₄H₄IN₃O₂
MDL Number: MFCD00210162
CAS Number: 13369-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR