3-Ethoxy-N,N-diethylaniline 97%

Supplier: Ambeed
Warning

Synonyms: N,N-Diethyl-m-phenetidine

A843606-1G A843606-25G A843606-5G
77281-668EA 15 USD
77281-668 77281-670 77281-672
3-Ethoxy-N,N-diethylaniline 97%
3-Ethoxy-N,N-diethylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO
MDL Number: MFCD00043936
CAS Number: 1864-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR