N-Butyl-3-methylpyridinium bis(trifluoromethylsulfonyl)imide 99%

Supplier: Ambeed
Warning

Synonyms: 1-Butyl-3-methylpyridin-1-ium bis((trifluoromethyl)sulphonyl)amide, BMPIm

A843391-100G A843391-1G A843391-25G A843391-5G
77244-468EA 174.07 USD
77244-468 77244-470 77244-472 77244-474
N-Butyl-3-methylpyridinium bis(trifluoromethylsulfonyl)imide 99%
N-Butyl-3-methylpyridinium bis(trifluoromethylsulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₂H₁₆F₆N₂O₄S₂
MW: 430.3869 g/mol
MDL Number: MFCD03788916
CAS Number: 344790-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR