Isoindolin-5-ol hydrochloride 97%

Supplier: Ambeed
Warning

A843361-100MG A843361-250MG
77244-464EA 56 USD
77244-464 77244-466
Isoindolin-5-ol hydrochloride 97%
Isoindolin-5-ol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
MDL Number: MFCD10700137
CAS Number: 1126832-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR