2-Bromo-1-indanol 99%

Supplier: Ambeed
Warning

A843286-100G A843286-25G A843286-500G A843286-5G
77268-540EA 109 USD
77268-540 77268-542 77268-544 77268-546
2-Bromo-1-indanol 99%
2-Bromo-1-indanol

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD00003798
CAS Number: 5400-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR