4-Fluorothioanisole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluorophenyl methyl sulphide, 1-Fluoro-4-(methylthio)benzene

A843079-100G A843079-1G A843079-25G A843079-5G
77293-970EA 63 USD
77293-970 77293-972 77293-974 77293-976
4-Fluorothioanisole 98%
4-Fluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₇FS
MDL Number: MFCD00040829
CAS Number: 371-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR