Tetrakis(acetonitrile)copper(I) tetrafluoroborate 97%

Supplier: Ambeed
Danger

Synonyms: Copper(I) tetra(acetonitrile) tetrafluoroborate, Tetrakis(acetonitrile)copper(I) tetrafluoroborate

A842358-1G A842358-25G A842358-5G A842358-10G
77244-458EA 12.36 USD
77244-458 77244-460 77244-462 77747-607
Tetrakis(acetonitrile)copper(I) tetrafluoroborate 97%
Tetrakis(acetonitrile)copper(I) tetrafluoroborate

Precaución: for Research Use Only

Formula: C₈H₁₂BCuF₄N₄
MW: 314.56 g/mol
MDL Number: MFCD09265110
CAS Number: 15418-29-8
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR