3,4-Dichloro-5-nitrobenzotrifluoride 97%

Supplier: Ambeed
Warning

A841129-100G A841129-1G A841129-25G
77281-662EA 295 USD
77281-662 77281-664 77281-666
3,4-Dichloro-5-nitrobenzotrifluoride 97%
3,4-Dichloro-5-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂Cl₂F₃NO₂
MDL Number: MFCD01862035
CAS Number: 657-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR