3,6-Di(pyridin-3-yl)-1,2,4,5-tetrazine 97%

Supplier: Ambeed
Warning

A840556-100MG A840556-1G A840556-250MG A840556-5G
77268-528EA 15.45 USD
77268-528 77268-530 77268-532 77268-534
3,6-Di(pyridin-3-yl)-1,2,4,5-tetrazine 97%
3,6-Di(pyridin-3-yl)-1,2,4,5-tetrazine

Precaución: for Research Use Only

Formula: C₁₂H₈N₆
MW: 236.23 g/mol
MDL Number: MFCD00234777
CAS Number: 107599-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR