4-Methoxyquinoline-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A840523-250MG A840523-5G A840523-1G
77293-962EA 15 USD
77293-962 77293-964 77749-642
4-Methoxyquinoline-2-carboxylic acid 97%
4-Methoxyquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MDL Number: MFCD00012336
CAS Number: 15733-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR