PLX-8394 98%

Supplier: Ambeed
Warning

Synonyms: (R)-N-(3-(5-(2-cyclopropylpyrimidin-5-yl)-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)-3-fluoropyrrolidine-1-sulphonamide, PLX8394

A840010-1MG A840010-25MG A840010-2MG A840010-50MG A840010-5MG A840010-250MG A840010-10MG A840010-100MG
77244-442EA 32.96 USD
77244-442 77244-444 77244-446 77244-448 77618-352 77747-604 77747-603 77747-602
PLX-8394 98%
PLX-8394

Precaución: for Research Use Only

Formula: C₂₅H₂₁F₃N₆O₃S
MW: 542.54 g/mol
MDL Number: MFCD29472263
CAS Number: 1393466-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR