Dicarbonylchloro[(1,2,3,4,5-η)-1,2,3,4,5-pentaphenyl-2,4-cyclopentadien-1-yl]-Ruthenium 98%
Supplier: Ambeed
|
Warning
|
A839553-100MG
A839553-1G
A839553-250MG
77268-522EA
88.58
USD
77268-522
77268-524
77268-526
Dicarbonylchloro[(1,2,3,4,5-η)-1,2,3,4,5-pentaphenyl-2,4-cyclopentadien-1-yl]-Ruthenium 98%
Dicarbonylchloro[(1,2,3,4,5-η)-1,2,3,4,5-pentaphenyl-2,4-cyclopentadien-1-yl]-Ruthenium
Precaución: for Research Use Only
Formula:
C₃₇H₂₅ClO₂Ru MW: 638.12 g/mol |
MDL Number:
MFCD09265041 CAS Number: 677736-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...