(S)-(-)-[(S)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane 98%

Supplier: Ambeed
Warning

Synonyms: (1Sp)-1-(Dicyclohexylphosphino)-2-[(S)-[2-(dicyclohexylphosphino)phenyl](dimethylamino)methyl]ferrocene

A839329-100MG A839329-250MG A839329-50MG A839329-1G
77256-222EA 92.7 USD
77256-222 77256-224 77256-226 77747-597
(S)-(-)-[(S)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane 98%
(S)-(-)-[(S)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane

Precaución: for Research Use Only

Formula: C₄₃H₆₃FeNP₂
MW: 711.76 g/mol
MDL Number: MFCD04117706
CAS Number: 914089-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR