4-Methoxy-1H-indole-2-carbaldehyde 97%

Supplier: Ambeed
Warning

A838720-100MG A838720-1G A838720-250MG
77244-426EA 99.91 USD
77244-426 77244-428 77244-430
4-Methoxy-1H-indole-2-carbaldehyde 97%
4-Methoxy-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.18 g/mol
MDL Number: MFCD13185674
CAS Number: 213682-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR