4-Benzoylphenyl methacrylate 98% stabilized

Supplier: Ambeed
Warning

Synonyms: 4-Methacryloyloxybenzophenone, Methacrylic acid 4-Benzoylphenyl ester

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A838608-25G A838608-5G
77268-518EA 45.32 USD
77268-518 77268-520
4-Benzoylphenyl methacrylate 98% stabilized
4-Benzoylphenyl methacrylate

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₃
MW: 266.30 g/mol
MDL Number: MFCD30470334
CAS Number: 56467-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR