(5aR,10bS)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride 98%
Supplier: Ambeed
|
Warning
|
A838366-100MG
A838366-1G
A838366-250MG
77256-216EA
63
USD
77256-216
77256-218
77256-220
(5aR,10bS)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride 98%
(5aR,10bS)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride
Precaución: for Research Use Only
Formula:
C₂₁H₂₂ClN₃O |
MDL Number:
MFCD09787522 CAS Number: 903571-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...