4-Amino-2-methyl-1-butanol 95%

Supplier: Ambeed
Danger

Synonyms: 4-Amino-2-methylbutan-1-ol

A838335-1G A838335-250MG
77293-952EA 19.57 USD
77293-952 77293-954
4-Amino-2-methyl-1-butanol 95%
4-Amino-2-methyl-1-butanol

Precaución: for Research Use Only

Formula: C₅H₁₃NO
MW: 103.17 g/mol
MDL Number: MFCD03093615
CAS Number: 44565-27-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR