1,3-Di-tert-butylimidazolium tetrafluoroborate 98%
Supplier: Ambeed
|
Warning
|
A838287-10G
A838287-1G
A838287-250MG
A838287-5G
77244-418EA
218
USD
77244-418
77244-420
77244-422
77244-424
1,3-Di-tert-butylimidazolium tetrafluoroborate 98%
1,3-Di-tert-butylimidazolium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₁₁H₂₁BF₄N₂ |
MDL Number:
MFCD07784425 CAS Number: 263163-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...