3-Methyl-2-buten-1-ol 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methyl-2-buten-4-ol

A837946-25G A837946-100G
77281-636EA 5.15 USD
77281-636 77749-637
3-Methyl-2-buten-1-ol 98%
3-Methyl-2-buten-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₀O
MW: 86.13 g/mol
MDL Number: MFCD00002916
CAS Number: 556-82-1
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR