PIK-III 98+%

Supplier: Ambeed
Warning

Synonyms: 4'-(Cyclopropylmethyl)-N2-(pyridin-4-yl)-[4,5'-bipyrimidine]-2,2'-diamine, Vps34-PIK-III

A837347-10MG A837347-1MG A837347-25MG A837347-50MG A837347-5MG
77268-508EA 33.99 USD
77268-508 77268-510 77268-512 77268-514 77268-516
PIK-III 98+%
PIK-III

Precaución: for Research Use Only

Formula: C₁₇H₁₇N₇
MW: 319.36 g/mol
MDL Number: MFCD28963904
CAS Number: 1383716-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR