2-Chlorophenethyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-(2-Bromoethyl)-2-chlorobenzene, 2-Chlorophenethyl Bromide, 2-(2-Chlorophenyl)ethyl bromide, 2-Chlorophenethyl bromide

A836925-10G A836925-250MG A836925-5G
77244-412EA 89 USD
77244-412 77244-414 77244-416
2-Chlorophenethyl bromide 97%
2-Chlorophenethyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MW: 219.51 g/mol
MDL Number: MFCD04117464
CAS Number: 16793-91-2
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR