3-Dimethylamino-1-pyridin-2-ylpropenone 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Dimethylamino)-1-(pyridin-2-yl)prop-2-en-1-one

A836582-10G A836582-1G A836582-25G
77281-624EA 45.32 USD
77281-624 77281-626 77281-628
3-Dimethylamino-1-pyridin-2-ylpropenone 97%
3-Dimethylamino-1-pyridin-2-ylpropenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
MDL Number: MFCD00115039
CAS Number: 66521-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR