5-Chloro-1H-indole-2-carboxamide 97%

Supplier: Ambeed
Warning

A836333-1G A836333-250MG
77268-504EA 27 USD
77268-504 77268-506
5-Chloro-1H-indole-2-carboxamide 97%
5-Chloro-1H-indole-2-carboxamide

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MDL Number: MFCD07364576
CAS Number: 21109-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR