4-Methyltetrahydro-2H-pyran-4-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A835786-1G
A835786-250MG
77268-500EA
297.67
USD
77268-500
77268-502
4-Methyltetrahydro-2H-pyran-4-carbonitrile 97%
4-Methyltetrahydro-2H-pyran-4-carbonitrile
Precaución: for Research Use Only
Formula:
C₇H₁₁NO MW: 125.168 g/mol |
MDL Number:
MFCD13180512 CAS Number: 856255-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...