(1R,1'R)-1,1',2,2',3,3',4,4'-Octahydro-1,1'-biisoquinoline ≥97%, ee 99%

Supplier: Ambeed
Warning

A835566-100MG A835566-1G A835566-250MG
77256-206EA 143 USD
77256-206 77256-208 77256-210
(1R,1'R)-1,1',2,2',3,3',4,4'-Octahydro-1,1'-biisoquinoline ≥97%, ee 99%
(1R,1'R)-1,1',2,2',3,3',4,4'-Octahydro-1,1'-biisoquinoline

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₂
CAS Number: 634180-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR