2-(4-(2-Methoxybenzamido)benzamido)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: EG1

A835001-10MG A835001-1MG A835001-25MG A835001-5MG
77268-492EA 28.84 USD
77268-492 77268-494 77268-496 77268-498
2-(4-(2-Methoxybenzamido)benzamido)benzoic acid 98%
2-(4-(2-Methoxybenzamido)benzamido)benzoic acid

Precaución: for Research Use Only

Formula: C₂₂H₁₈N₂O₅
MW: 390.38 g/mol
MDL Number: MFCD03624354
CAS Number: 693241-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR