4'-Chloro-3'-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-nitroacetophenone

A834344-100G A834344-25G A834344-5G
77281-612EA 108 USD
77281-612 77281-614 77281-616
4'-Chloro-3'-nitroacetophenone 98%
4'-Chloro-3'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClNO₃
MDL Number: MFCD00007083
CAS Number: 5465-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR