6,6'-(6-(4-Methoxyphenyl)-1,3,5-triazine-2,4-diyl)bis(3-((2-ethylhexyl)oxy)phenol) 97%

Supplier: Ambeed
Warning

A834261-5G A834261-25G A834261-100G
77268-486EA 7 USD
77268-486 77749-629 77749-628
6,6'-(6-(4-Methoxyphenyl)-1,3,5-triazine-2,4-diyl)bis(3-((2-ethylhexyl)oxy)phenol) 97%
6,6'-(6-(4-Methoxyphenyl)-1,3,5-triazine-2,4-diyl)bis(3-((2-ethylhexyl)oxy)phenol)

Precaución: for Research Use Only

Formula: C₃₈H₄₉N₃O₅
MDL Number: MFCD08435974
CAS Number: 187393-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR