(±)-2-Bromopropionyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromopropionylchloride

A834224-25G A834224-5G
77268-482EA 54 USD
77268-482 77268-484
(±)-2-Bromopropionyl chloride 98%
(±)-2-Bromopropionyl chloride

Precaución: for Research Use Only

Formula: C₃H₄BrClO
MW: 171.42
MDL Number: MFCD00000711
CAS Number: 7148-74-5
UN: 2924
ADR: 3,8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR