2-Hydroxy-5-(trifluoromethyl)nicotinaldehyde 95%

Supplier: Ambeed
Warning

A833623-250MG A833623-50MG
77268-478EA 206 USD
77268-478 77268-480
2-Hydroxy-5-(trifluoromethyl)nicotinaldehyde 95%
2-Hydroxy-5-(trifluoromethyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₂
MW: 191.10 g/mol
MDL Number: MFCD09909904
CAS Number: 129904-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR