Vanillin oxime 97%

Supplier: Ambeed
Danger

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde oxime

A833423-1G A833423-5G
77256-202EA 27.81 USD
77256-202 77256-204
Vanillin oxime 97%
Vanillin oxime

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD00019965
CAS Number: 2874-33-1
UN: 2926
ADR: 4.1,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR